Recirculating coolers, FL series
Supplier: JULABO GmbH
9661006 9661012 9663012 9660003 9661017 9663017 9663025 9663040 9666025 9666040 9666070 9666110 9671017 9673017 9673025 9673040 9676025 9676040 9676070 9676110 9666200 9676200
461-3466EA 4865 EUR
461-3466 461-3467 461-3469 461-3465 461-3468 461-3470 461-3471 461-3472 461-3473 461-3474 461-3475 461-3476 461-3477 461-3478 461-3479 461-3480 461-3481 461-3482 461-3483 461-3484 461-0638 461-0639
Recirculating coolers, FL series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Compact, ergonomically designed chillers which have integrated stainless steel tanks with large volumes. Models can be either air or water cooled. Applications include use with rotary evaporators, autoclaves, reaction vessels and for routine laboratory cooling. All units have an LED temperature display for actual/set point integrated into a splash-proof keypad and recessed grips for easy relocation integrated in the housing. A hinged tray serves as a compartment for the operating manual and other documents. The removable venting grid allows for easy cleaning of the condenser. Venting slotsMore Product Information