Drying ovens, Heratherm® General protocol series
Supplier: Thermo Fisher Scientific
51028148 51028149 51028150 51028139 51028140 51028141
466-0264EA 2490 EUR
466-0264 466-0265 466-0266 466-0261 466-0262 466-0263
Drying ovens, Heratherm® General protocol series
Ovens Standard Ovens
Heating and drying ovens with stainless steel chambers, which are designed for routine applications. Units with natural convection are ideal for gentle drying and heating, to protect delicate samples. Units with forced convection are suitable for faster drying and heating, and superior temperature uniformity.
  • Automatic over-temperature alarm system
  • Built-in timer
  • Large, easy to read vacuum fluorescent display and microprocessor-controlled, touch-button operation
  • Doors can be opened over 180° to allow easy access to the interior
  • OGS - exhaust vent can be