Drying ovens, Heratherm® advanced protocol security series
Supplier: Thermo Fisher Scientific
51028154 51028155 51028156 51028545 51028145 51028146 51028147 51028546 51028547 51028548 51028549 51028544
466-0285EA 3460 EUR
466-0285 466-0286 466-0287 466-0283 466-0279 466-0280 466-0281 466-0284 466-0288 466-0289 466-0290 466-0282
Drying ovens, Heratherm® advanced protocol security series
Ovens Standard Ovens
Heating and drying ovens with coated or stainless steel exterior. The timer allows the oven to be turned on or off after a preset time, with a choice between weekly, hourly or real time settings. Units with natural convection are ideal for gentle drying and heating, to protect delicate samples. Units with forced convection have adjustable fan speed and are suitable for faster drying and heating, and superior temperature uniformity.
  • Standard over-temperature alarm and additional under-temperature alarm
  • Large, easy to read vacuum fluorescent display and