Desiccator cabinets, Lab Companion™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F424004001 F424004131 F424004111 F424004021 F424004121 F424004011 F424004031 F424004101
467-0141EA 3140 EUR
467-0141 467-0146 467-0144 467-0142 BELAF424004121 467-0143 467-0145 BELAF424004101
Desiccator cabinets, Lab Companion™, SP Bel-Art
Desiccators Desiccator Cabinets
These compact, high-quality vacuum desiccators with clear or amber (UV blocking) polycarbonate maintain a vacuum and stay gastight allowing reliable dry storage or experiments over extended periods.
  • Stackable and provide vacuum integrity resulting in lower maintenance
  • Able to maintain 29 inches Hg vacuum for over 72 hours at room temperature
  • Single piece moulded design with no leakage and exceptional durability
  • Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release or gas exchange
  • Easy to clean, chemical resistant and allow quick