Laboratory freezers, FROSTER-LABEX®-530/-730
Supplier: KIRSCH PHILIPP
10331 10332
471-0417EA 8490 EUR
471-0417 471-0418
Laboratory freezers, FROSTER-LABEX®-530/-730
Freezers Laboratory / General Purpose Freezers
These laboratory freezers are designed with explosion-proof interior (intrinsically safe), re-circulating air cooling, automatic fast defrosting and a RS485 interface.
  • Self-closing door with right hinged
  • Micro-processor controlled temperature and thawing regulation to maximize temperature stability
  • Membrane keyboard with digital temperature display and minimum/maximum temperature memory
  • Control and display sensors protected by safety barriers
  • Alarms can be forwarded using a potential-free contact
  • Extra-thick 70 mm energy saving insulation
  • Door frame heater