2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 90%

Supplier: Thermo Fisher Scientific

474500010 474500050
474500010.EA 226 EUR
474500010. 474500050.
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 90%
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid)
Formula: C₁₈H₂₀N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00004247
CAS Number: 1170-02-1
EINECS: 214-625-3

Order Now

Specification Test Results

Appearance white to light brown crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR