Cryoboxes, 133×133 mm
Supplier: RATIOLAB
5120165 5131321 5132321 5131325 5132325 5131322 5132322 5131324 5132324 5131323 5132323 5131501 5132501 5131505 5132505 5131502 5132502 5131504 5132504 5131503 5132503 5131751 5132751 5131755 5132755 5131752 5132752 5131754 5132754 5131753 5132753 5131001 5132001 5131005 5132005 5131002 5132002 5131004 5132004 5131003 5132003 5131131 5132131 5131135 5132135 5131132 5132132 5131134 5132134 5131133 5132133 5120204 5120194 5120185 5120154 5120144 5120134 5120124 5120205 5120195 5120184 5120155 5120135 5120125 5120206 5120196 5120186 5120167 5120136 5120126 5120207 5120197 5120187 5120137 5120127 5120214 5120215 5120216 5120217 5120225 5120226 5120147
479-0154EA 2 EUR
479-0154 479-0300 479-0301 479-0302 479-0303 479-0304 479-0305 479-0306 479-0307 479-0308 479-0309 479-0310 479-0311 479-0312 479-0313 479-0314 479-0315 479-0316 479-0317 479-0318 479-0319 479-0320 479-0321 479-0322 479-0323 479-0324 479-0325 479-0326 479-0327 479-0328 479-0329 479-0330 479-0331 479-0332 479-0333 479-0334 479-0335 479-0336 479-0337 479-0338 479-0339 479-0340 479-0341 479-0342 479-0343 479-0344 479-0345 479-0346 479-0347 479-0348 479-0349 479-0350 479-0351 479-0352 479-0355 479-0356 479-0357 479-0358 479-0360 479-0361 479-0362 479-0365 479-0367 479-0368 479-0370 479-0371 479-0372 479-0374 479-0377 479-0378 479-0380 479-0381 479-0382 479-0387 479-0388 RATI5120214 RATI5120215 RATI5120216 RATI5120217 RATI5120225 RATI5120226 RATI5120147
Cryoboxes, 133×133 mm
Boxes Cryoboxes
Cryoboxes, cryo resistant cardboard.
  • Various combinations possible with different grid inserts
  • Height stated is the interior height
  • Supplied with lid
For storage of cryovials at low temperatures.
Dimensions: 133 x 133 mm
Height stated is the interior height
Ordering information: Grid insert to be ordered separately.