VWR® Labtop Coolers
Supplier: VWR Collection
479-0641EA 176 EUR
479-0641 479-0642 479-0643
VWR® Labtop Coolers
Boxes Cooling/Freezing Boxes and Bags
−20 °C mini labtop cooler PC, non toxic gel: The −20 °C mini labtop cooler holds 12×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over one hour. Ideal for bench top use or transport. Filled with non toxic gel. Prior to use, place in −25 °C freezer for at least 24 hours, store at −25 °C between use.
−20 °C maxi labtop cooler PC, non toxic gel
The −20 °C maxi labtop cooler, holds 32×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over 1 hour. Suitable for bench top use or transport. Wire handle secures top while carrying. Cooler and lid filled with non toxic gel.More Product Information