Racks for cryoboxes, standard type
Supplier: TENAK
TE23120 TE23122 TE76010 TE23156 TE23190 TE23186 TE23184 TE23182 TE23180 TE23174 TE23172 TE23194 TE23160 TE23200 TE23154 TE23152 TE23150 TE23138 TE23136 TE23134 TE23132 TE23130 TE23124 TE23170 TE76020 TE23192 TE23222 TE23220 TE23212 TE23210 TE23050 TE23052 TE23054 TE23056 TE23060 TE23062 TE23064 TE23066 TE23068 TE23070 TE23080 TE23082 TE23084 TE23086 TE23230 TE23232 TE23240 TE23242 TE23244 TE23250 TE23252 TE23132B TE23134B TE23136B TE23122B TE23124B TE23130B TE23138B TE23150B TE23152B TE23154B TE23156B TE23160B TE23170B TE23172B TE23174B TE23180B TE23182B TE23184B TE23186B TE23190B TE23192B TE23194B TE23200B TE23210B TE23212B TE23220B TE23222B TE23334 TE23328 TE23330 TE23332 TE23336 TE23338 TE23154-S
479-0730EA 103 EUR
479-0730 479-0731 479-0850 479-0741 479-0750 479-0749 479-0748 479-0747 479-0746 479-0745 479-0744 479-0752 479-0742 479-0753 479-0740 479-0739 479-0738 479-0737 479-0736 479-0735 479-0734 479-0733 479-0732 479-0743 479-0851 479-0751 479-0757 479-0756 479-0755 479-0754 479-1376 479-1715 479-1716 479-1717 479-1718 479-1719 479-1720 479-1721 479-1722 479-1723 479-1724 479-1725 479-1726 479-1727 479-1752 479-1753 479-1754 479-1755 479-1756 479-1757 479-1758 479-1003 479-1730 479-1731 479-1002 479-1728 479-1729 479-1732 479-1733 479-1004 479-1734 479-1735 479-1736 479-1737 479-1738 479-1739 479-1740 479-1741 479-1742 479-1743 479-1744 479-1745 479-1746 479-1747 479-1748 479-1749 479-1750 479-1751 479-1318 479-1315 479-1316 479-1317 479-1319 479-1320 479-1870
Racks for cryoboxes, standard type
Racks Cryogenic Box Racks
Stainless steel.
  • Racks for five different box heights
  • Can be used with all freezer brands
  • Fits all standard cryoboxes
For upright freezers. These racks are an inexpensive storage system. They will fit into any freezer, and can be used with all standard cryoboxes. Measure the inside height and depth of your freezer before choosing the right rack.
Stainless steel safety spring lock
Can be used when there is a gap between the cryobox and the shelf from 1 to 6 mm
For Tenak classic and budget racks, or any other rack brand with a similar design