Cryogenic vials, Nalgene®
Supplier: Thermo Fisher Scientific
5000-0012 5000-0020 5000-0050 5000-1012 5000-1020 5005-0015 5011-0012 5011-0020 5012-0012 5012-0020
479-3221EA 208 EUR
479-3221 479-3222 479-3223 479-3224 479-3225 479-0006 479-0007 479-0008 479-0009 479-0010
Cryogenic vials, Nalgene®
Vials
These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to the vapour phase of liquid nitrogen (at temperatures as low as –150 °C). They are made from PPCO and have HDPE screw caps.
  • External thread
  • Self standing
  • Non cytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
  • Tubes with writing area are graduated
System 100™ tubes allow for maximum storage capacity and have a PP closure and internal silicone gasket.