Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
363401 363452 366656 368632 375418 377267 351934 337516 347597 347627 347643 366524 375353 377224 379146 379189 340711 367997 363452_CZ
479-6837EA 206 EUR
479-6837 479-6839 479-6840 479-6841 479-6847 479-6843 479-6852 479-6848 479-6849 479-6850 479-6836 479-6838 479-6846 479-6842 479-6845 479-6844 479-6853 479-1159 479-3202
Cryogenic tubes, CryoTubes™, Nunc™
Vials
PP tubes with PP screw cap (internal thread vials) or PE screw cap (external thread vials) and white writing area, sterile. Developed for storage of cell cultures, bacteria and other samples at extremely low temperatures. Suitable for use in freezers as well as in the vapour phase of liquid nitrogen*.
Non toxic, non pyrogenic.
Conform to IATA regulations for the air transportation of biological samples.
* Warning: Do not use CryoTubes™ in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ tubing. Improper use may cause entrapment of liquefiedMore Product Information