Centrifugal filtration units, Vivaspin® Turbo 15
Supplier: Sartorius
VS15T01 VS15T02 VS15T11 VS15T12 VS15T21 VS15T22 VS15T31 VS15T32 VS15T41 VS15T42 VS15T91 VS15T92
512-0557EA 174 EUR
512-0557 512-0558 512-0559 512-0560 512-0561 512-0562 512-0563 512-0564 512-0565 512-0566 512-0567 512-0568
Centrifugal filtration units, Vivaspin® Turbo 15
Filters Centrifugal Filters
Vivaspin® Turbo 15 can handle 0,1 to 15 ml sample volume in swing bucket rotors and 11 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
  • High retentate recovery >95%
  • pH 1 to 14 suited PS housing with high chemical compatibility
  • Safe sample concentration and easy recovery due to unique, angular and pipette friendly dead-stop pocket
  • Volume graduation for easy detection/monitoring of the concentrate volume
The Vivaspin® Turbo 15 optimised design, the sleek internal profile, ensures maximum process speeds right the way down to the last fewMore Product Information