Anopore inorganic membranes without support ring, Whatman™
Supplier: Whatman products (Cytiva)
6809-7003 6809-7013 6809-7023 6809-5502 6809-5522
514-0521EA 741 EUR
514-0521 514-0522 514-0523 514-0662 514-0663
Anopore inorganic membranes without support ring, Whatman™
Filters Membrane Filters
Whatman Anopore inorganic membranes, supplied as Anodisc membrane filters, are an aluminum oxide membrane with a honeycomb pore structure that facilitates highly precise filtration. Their transparency and surface retention enable efficient light or electron microscopy.
  • Virtually transparent when wet
  • Minimal autofluorescence
  • Hydrophilic with wide solvent compatibility
  • Non-toxic for cellular growth
The non-deformable, capillary pore structure has no lateral crossovers between individual pores. The inorganic membrane filters have high pore density andMore Product Information