Syringe filters, CA, Puradisc™, Whatman™
Supplier: Whatman products (Cytiva)
6771-1304 10462100 10462200 10462205 10462206 10462240 10462241 10462243 10462260 10462261 10462263 10462600 10462601 10462610 10462700 10462701 10462710
516-0924EA 262 EUR
516-0924 514-1113 514-1111 514-1112 514-1122 514-1114 514-1124 516-0909 514-1115 514-1125 514-1120 514-1123 514-1127 514-1140 514-1121 514-1117 514-1141
Syringe filters, CA, Puradisc™, Whatman™
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • From the Puradisc FP 30 range
  • Eight different material options
  • Choice of 4 mm, 13 mm, 25mm and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters caterMore Product Information