Phase separating paper, grade 1PS
Supplier: Whatman products (Cytiva)
2200-070 2200-090 2200-110 2200-125 2200-150 2200-185 2200-240 2200-270 2200-654
516-5201EA 50.8 EUR
516-5201 515-0086 515-0087 515-0088 515-0089 515-0090 515-5017 515-5018 WHAT2200-654
Phase separating paper, grade 1PS
Filters Filter Papers Phase Separation Paper
1PS phase-separating paper is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
  • Impregnated with stabilised silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the 1PS phase-separating paper. The solvent phase flows throughMore Product Information