Fixed-angle rotor package, MLA-130
Supplier: Beckman Coulter
367114
521-4043EA 17160 EUR
521-4043
Fixed-angle rotor package, MLA-130
Centrifuges Rotors
Major applications: Rapid differential sedimentation (pelleting) of small particles such as subcellular organelles and viruses.