VWR® Centrifuge Tubes, Conical Bottom, Flip Cap
Supplier: VWR Collection
525-1047EA 213 EUR
525-1047 525-1048 525-1045 525-1046
VWR® Centrifuge Tubes, Conical Bottom, Flip Cap
Tubes Centrifuge Tubes
Centrifuge tubes are ideal for use in centrifuging samples or storage in many research areas.
  • Manufactured from high grade polypropylene
  • Easy flip-open caps enable single-handed access
  • Autoclavable at 121 °C, and freezable to −80 °C
  • Sterilised by electron beam irradiation, DNase-free, RNase-free and non pyrogenic
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Tubes feature black printed graduations and writing area can withstand chloroform.
Certifications: More Product Information