Chloroform stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: ∼ 1% Stabilised with ethanol

5285.2500EA 282.17 EUR
5285.2500
Chloroform stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1 III
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99.8 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Preservative (Ethanol) about 1%
Water (KF) max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR