SpeedVac™ SPD1030P1 Integrated Vacuum Concentrator System Kit
Supplier: THERMO SAVANT
SPD1030P1-230
531-1494EA 18000 EUR
531-1494
SpeedVac™ SPD1030P1 Integrated Vacuum Concentrator System Kit
Concentrators
Thermo Scientific™ Savant™ SpeedVac™ SPD1030 and SPD2030 vacuum concentrators are fully integrated systems that combine a concentrator, refrigerated cold trap, diaphragm pump and a vacuum gauge into a single, compact unit.
  • Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
  • Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
  • Ease of use: Intuitive design with a bright display, three preset and three