Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

5329.6025EA 0 EUR
5329.6025
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Titer 1.185 - 1.215 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR