VWR®, Screw Neck Vials ND10
Supplier: VWR Collection
548-0386AEA 286 EUR
548-0386A 548-2499A 548-0385A 548-0387A 548-3281A 548-3187A 548-1418A 548-3260A 548-1397A 548-2589A 548-2590A
VWR®, Screw Neck Vials ND10
Vials
ND10 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
ND10 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with glass or plastic inserts.
Associated screw caps mostly available with assembled septa are engineered toMore Product Information