VWR®, Polypropylene Microplates, 96-Well
Supplier: VWR Collection
548-3375EA 72 EUR
548-3375
VWR®, Polypropylene Microplates, 96-Well
Microplates Deep Well Microplates
VWR® 96-Well plates and covers are designed for chromatography and high throughput screening (HTS) applications.
  • SBS and ANSI standard footprint
  • Low binding virgin PP for homogenous assays and storage
  • –80 to +121 °C tolerant
VWR® 96-Well Microplates are manufactured specifically for chromatography. Multiple volumes and interior finishes allow for enhanced sample recovery. Low binding virgin polypropylene plates are perfect for high throughput screening (HTS), and sensitive samples.