Solid phase extraction cartridges, Bond Elut™ C8
Supplier: VARIAN
12162029B 12166002B 12256024 75403025 12102029 12102002 12102026 12102053 12113002 12113028 12102059 7540301C 12102100 12256002 12256032 12105028 12113025 12113075 7550301C 75403050 7540302C 52102002 52102026
551-0374EA 0 EUR
551-0374 551-0381 549-0825 551-0428 551-0316 551-0301 551-0313 549-0786 VARI12113002 VARI12113028 549-0788 551-0427 549-0798 549-0820 549-0827 549-0807 VARI12113025 VARI12113075 VARI7550301C VARI75403050 VARI7540302C 549-0872 549-0873
Solid phase extraction cartridges, Bond Elut™ C8
Chromatography Columns
Bond Elut™ C8 is very similar in properties to C18, but is not as retentive for non-polar compounds, due to its shorter hydrocarbon chain, and therefore reduced carbon loading. C8 is an excellent replacement for C18 when analytes are too strongly retained for effective elution. The potential for polar interactions is somewhat higher than for C18 because there is less coverage of the silica surface. These polar interactions are not, however, a significant property of C8.
  • Excellent for strongly-retained analytes
  • Polar interactions not significant
  • Less retentive than C18