Chromatographic columns with frit, PTFE or valve stopcock
Supplier: Lenz Laborglas GmbH & CO.KG
05450701 05450702 05450703 05450704 05450706 05450708 05445704 05445706 05445708 05450800 05450804 05445701 05445702 05450700 05450801 05450802 05450803 05450806
552-5401EA 76 EUR
552-5401 201-0744 201-0745 201-0593 201-0746 201-0747 201-0592 201-0741 201-0742 201-0748 201-0752 201-0739 201-0740 201-0743 201-0749 201-0750 201-0751 201-0753
Chromatographic columns with frit, PTFE or valve stopcock
Chromatography Columns
These chromatography columns are made from DURAN® tubing.
  • Columns with sintered frit - porosity zero