Waste Tubing Desktop and Rear Mounts, SymLine®
Supplier: S.C.A.T.
106400 106450 106407 106412 106612 106455 106601 106616
554-1693EA 118.6 EUR
554-1693 554-1694 554-4357 SCTV106412 SCTV106612 SCTV106455 SCTV106601 SCTV106616
Waste Tubing Desktop and Rear Mounts, SymLine®
Tubing Transfer Tubing
The filling units are mounted in space-saving fashion within the workspace. Available as a special HPLC connection, for direct connection to waste tubing.
  • Desktop mount can be integrated into nearly all working areas
  • Flat desktop mount and HPLC waste collector in one
  • Rear wall mount uses available working area capacity to provide more space for laboratory samples and experimental setups
  • Waste can be disposed directly in the fume cupboard
The desktop mount can be integrated into nearly all working areas. Whether used in conjunction with a work table orMore Product Information