Gas purifiers, GateKeeper® GPU 700 4R
Supplier: ENTEGRIS
GPUS700FIX04R0 GPUS700FHX04R0 GPUS700FOP04R0
568-0922EA 0 EUR
568-0922 568-0921 568-0923
Gas purifiers, GateKeeper® GPU 700 4R
Filters Gas Filters
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 700 is designed for peak flow rates up to 120 slpm.