Anti-Foxp3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 3G3]
Supplier: Tonbo Biosciences
60-5773-U025 60-5773-U100
60-5773-U025EA 210 EUR
60-5773-U025 60-5773-U100
Anti-Foxp3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 3G3]
Antibodies
The 3G3 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is expressed at constitutively high levels in Treg cells, which are further identified as being CD4+ CD25+. In resting conventional T cells (CD4+ CD25-) Foxp3 expression is restricted, and upon TCR activation is expressed only transiently and in a small proportion of cells. However, the growth factor TGF-beta has been shown to induce expression of Foxp3 in naïve T cells, driving theirMore Product Information