Texas Red®-DHPE stain_membrane, Histology

Supplier: Biotium

Synonyms: (N-(Texas Red® sulphonyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt

60027
60027.EA 306 EUR
60027.
Texas Red®-DHPE stain_membrane, Histology
Texas Red®-DHPE

Texas Red®-DHPE is a rhodamine-labeled glycerophosphoethanolamine lipid.


  • λEx/λEm (MeOH) = 582/601 nm
  • Dark solid soluble in chloroform
  • Store at -20°C and protect from light


DHPE conjugates of rhodamine dyes have been used as fluorescence energy acceptors in combination with NBDPE as the donor in membrane fusion assays using the principle of fluorescence energy transfer (FRET). These probes have also been used for following membrane trafficking during endocytosis.

Formula: C₇₄H₁₁₇N₄O₁₄PS₂
MW: 1380.77 g/mol
Storage Temperature: Freezer
CAS Number: 187099-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=541273be83c546199381c4b559ae6566ccd655b4&vid=541273be83c546199381c4b559ae6566ccd655b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR