Semi-micro and analytical balances, Excellence Plus XP series
Supplier: Mettler - Toledo
11106047 11106027 11106024 11106026 11106029 11106030 11106032 11106036 11106038 11106042 11106044 11106045 11130052 11130054 11130076 11130078 11130082 11130084
611-2024EA 6770 EUR
611-2024 611-2015 611-2013 611-2014 611-2016 611-2017 611-2018 611-2019 611-2020 611-2021 611-2022 611-2023 611-2092 611-2093 611-2025 611-2026 611-2027 611-2028
Semi-micro and analytical balances, Excellence Plus XP series
Balances and Scales
The XP analytical balances provide high performance weighing with improved linearity safety and security. This is achieved with the introduction of QM toolbox, SmartSens and MinWeigh functions alongside the SmartGrid technology, which provide fast and accurate weighing. This range is therefore particularly suited where regulatory compliance is required. The draught shield and drip tray can be completely, quickly and easily removed as before but now SmartSens allows the operator to weigh without touching the balance, thereby smoothing the work-flow and reducing spillage and contaminationMore Product Information