Industrial scales, Excellence Plus XP series
Supplier: Mettler - Toledo
11130233 11130239 11130245 11130251 11130257 11130263 11130630 11130632 11130635 11130636 11130638 11130639 11130641 11130642 11130644 11130645 11130648 11130651 11130735 11130738 11130741 11130633 11130647 11130650 11130653 11130732
611-2048EA 4210 EUR
611-2048 611-2050 611-2052 611-2054 611-2056 611-2058 611-2062 611-2063 611-2064 611-2065 611-2066 611-2067 611-2068 611-2069 611-2070 611-2071 611-2072 611-2073 611-2084 611-2086 611-2088 611-2146 611-2147 611-2148 611-2149 611-2152
Industrial scales, Excellence Plus XP series
Balances and Scales
The large platform versions of XP balances provide fast, stable results, safety and security, making the range particularly suitable where regulatory compliance is required for high capacity weighing. The design features MonoBloc HighSpeed technology, a large, colour, backlit touch screen that facilitates direct access to all functions and an extended range of applications that include density determinations, formulation, piece counting, statistics and dynamic weighing. Commercial keyboards or barcode readers can be easily connected with the PS/2 interface option or connectivity can beMore Product Information