VWR® SMG Advanced Semi-Micro Balances, Graphic Display
Supplier: VWR Collection
611-3551EA 1790 EUR
611-3551 611-3554 611-3557 611-3558 611-3561 611-3564 611-4952 611-4953 611-4954 611-4955
VWR® SMG Advanced Semi-Micro Balances, Graphic Display
Balances and Scales
Semi-micro balances with resolution of 0,01 mg, they are equipped with a new highly integrated electromagnetic cell. All models feature automatic internal calibration and are characterised by a high level of precision and repeatability. These VWR® semi-micro balances provide an excellent price/performance ratio, thanks to continuous intensive research and study of how weighing works. All models have a stainless steel top pan.
  • Integrated weighing sensor for enhanced stability and quick response time
  • Selectable filter levels and measuring units
  • Check