Precision balances, Adventurer®
Supplier: OHAUS
30100628 30100629 30100627 30100626 30100625 30100624 30100623 30127968 30127967 30127966 30127963 30127962 30127960 30122618 30122617 30122616 30122615 30122614 30127969 30127961 30127971 30206080 30206079 30206082 30206081 30206084 30206083 30127970 30122620 30122619 30122651
611-3797EA 2570 EUR
611-3797 611-3798 611-3796 611-3795 611-3794 611-3793 611-3792 611-3231 611-3230 611-3229 611-3226 611-3225 611-3223 611-3221 611-3220 611-3219 611-3218 611-3217 611-3232 611-3224 611-3723 611-3724 611-3565 611-3725 611-3566 611-3726 611-3567 611-3568 611-3721 611-3569 611-3722
Precision balances, Adventurer®
Balances and Scales
A range of balances with multiple features, but uncomplicated weighing capabilities, provide the user with rapid, accurate results for routine weighing and measurement activities. Applications include weighing, formulation, dynamic weighing, parts counting, totalisation, density determination, check weighing and percentage weighing. Stabilisation times are typically less than two seconds.
  • Stability, accuracy and fast operation
  • Wide viewing angle colour touch screen (109 mm) for quick access to nine application modes
  • Dual USB ports (front and rear) with