VWR® TS Professional Semi-micro Balances
Supplier: VWR Collection
611-3812EA 3060 EUR
611-3812 611-3813 611-3814 611-3811
VWR® TS Professional Semi-micro Balances
Balances and Scales
These semi-micro balances with 5" colour touch screen display and built-in ioniser ensure accurate sample preparation. Static charges accumulate in different samples that are routinely weighed in the laboratory. These static charges can build up and alter the weighing value. By blowing ions, the built-in ioniser neutralises static electricity and within seconds the sample is ready to be weighed. The balance calibration system automatically calibrates the balance when there is a temperature change or at a set period of time (factory settings), this ensures accurate measurements even if theMore Product Information