VWR® Safety Bottle Carriers
Supplier: VWR Collection
611-5280EA 58.5 EUR
611-5280 611-5281 611-5282 611-5283 611-5284 611-5285 611-5286 611-5287
VWR® Safety Bottle Carriers
Carriers Bottle Carriers/Containers
The safety bottle carrier provides a safe and reliable way to transport a variety of bottled reagents in the lab. Available in 1,5 and 4,5 L capacities, this carrier is the practical answer for any clinical and research lab setting.
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilises bottle in the centre of carrier for safer, steadier transport
  • Carriers come in four colours with designated labelling areas for easy