Weighing paper, grade MN 226
Supplier: MACHEREY-NAGEL
186002
611-9091EA 14 EUR
611-9091
Weighing paper, grade MN 226
Weighing Papers
This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that the weighed goods can be transferred without loss.