Inoculating loops, Corning® Gosselin™
Supplier: Corning
OS10-02 OS1-01 OS1B-01 OS10B-02
612-3691EA 322 EUR
612-3691 612-3692 GOSSOS1B-01 GOSSOS10B-02
Inoculating loops, Corning® Gosselin™
Spreaders Inoculating Loops and Needles
These polystyrene inoculating loops are highly ergonomic and are color coded for easy identification.
  • Spherical or needle extremity
  • Assured sterile 10-3
Traceability information is provided on each bag.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Borre