VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
612-4466EA 19.3 EUR
612-4466 612-4518 612-4468 612-4469 612-4470 612-4471 612-4472 612-4473 612-4520 612-4476 612-4479 612-4481 612-4486 612-4490 612-4491 612-4492 612-4494 612-4495 612-4496 612-4497 612-4499 612-4500 612-4502 612-4543 612-4504 612-4505 612-4506 612-4508 612-4509 612-4511 612-4512 612-4513 612-4514 612-4515 612-4542 612-4533 612-4526 612-4527 612-4530 612-4523 612-4536 612-4538 612-4545 612-3399 612-3608 612-3609 612-3610 612-3619 612-3627 612-5770 612-5781
VWR® Standard Line Transfer Pipettes
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Non toxic and inert to biological fluids and most acids
  • Can be sealed, refrigerated and gas sterilised
  • No bulb to insert or remove
  • Uniform drop size
The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
Ordering information: To help you find the correct product andMore Product Information