Bottle-top dispensers, seripettor® / seripettor® pro
Supplier: Brand
4720140 4720150 4720450 4720440 4720420 4720120
612-5625EA 169 EUR
612-5625 612-5626 612-3218 612-3217 613-1649 613-1648
Bottle-top dispensers, seripettor® / seripettor® pro
Bottle Top Dispensers
Variable volume.
  • Easily replaceable cartridge and seal
  • Automatic dispensing: Spring-loaded piston refills the cylinder after use
  • Volume setting: Indented track for quick and exact volume setting
  • Can be taken apart without tools, easy cleaning and maintenance
  • seripettor® for alkaline, buffer solutions and culture media, e.g. agar nutrient medium, up to +60 °C
  • seripettor® pro for dispensing many acidic, alkaline and saline solutions, as well as solvents such as essential oils
The seripettor® and seripettor® pro with innovative and economic design areMore Product Information