VWR® Reagent Reservoirs
Supplier: VWR Collection
613-1172EA 152 EUR
613-1172 613-1173 613-1186 613-1174 613-1175 613-1176 613-1177 613-1178 613-1179 613-1180 613-1181 613-1182 613-1183 613-1171 613-1185 613-1187 613-1184
VWR® Reagent Reservoirs
Reagent Reservoirs
Designed for use in multi-channel pipetting applications, these disposable reservoirs accommodate all types of pipettes up to 12-channel width.
  • Trough design, graduation marks on interior walls, and pour-off spouts on each corner
  • Lot certified, non pyrogenic, DNase- and RNase-free
  • Non sterile or electron beam sterilised
Ordering information: 25 ml reservoirs also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity; 50 ml reservoirs available in clear FDA-grade PVC or virgin-More Product Information