Microlab® 600 Syringe Dispensers
Supplier: HAMILTON BONADUZ
ML620-DIS ML635 ML615-DIS ML625-DIS ML630 ML610-DIS ML615-CNT
613-1567EA 7060 EUR
613-1567 613-1569 615-0025 613-2970 613-2971 615-0023 615-0024
Microlab® 600 Syringe Dispensers
Syringe Dispensing Systems
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
  • Dispense volumes from 100 µl to 50 ml
  • Inert fluid path compatible with harsh chemicals
The single syringe dispenserMore Product Information