Digital burettes, solarus®
Supplier: HIRSCHMANN
9392020 9392050 9392010
613-3255EA 1048 EUR
613-3255 613-3256 613-1476
Digital burettes, solarus®
Burettes
Variable volume.
  • Integral solar panel provides energy for electronic components such as the digital display, so batteries no longer need to be replaced or charged
  • Easy to use recirculation system for rapid priming with no loss of reagents
  • Bi-directional RS232 interface and USB
  • Parts that come into contact with liquids are autoclavable
  • Certificate of quality and conformity
These innovative digital burettes, with integrated photovoltaic cells and manual control, are designed for exact titration.
Ordering information: Supplied with three adaptersMore Product Information