BD Microlance™ 3 Needles
Supplier: BD Medical
300400 300635 300800 301155 301156 304000 304432 300637 300600 304434 301000UK 304000UK 303273 303262 310205
613-4952EA 12.2 EUR
613-4952 613-3832 613-3924 613-3934 613-3935 613-3942 613-4970 613-3897 613-0902 615-0034 613-1688 BDAM304000UK BDAM303273 BDAM303262 BDAM310205
BD Microlance™ 3 Needles
Syringe Needles
The BD Microlance™ 3 needles are designed for easy penetration, smooth gliding force during insertion/withdraw, and reduced cannula friction while sliding through tissue. These needles are available in a wide range of gauges, lengths, and bevel designs.
  • Recommended use: intradermal, subcutaneous, intramuscular, intravenous injections, and aspiration
  • BD Microlance™ needles can be connected to a luer slip adapter or luer-lock connector
  • Natural rubber latex is not part of the material formulation
  • Sterile, single-use design
Conventional needles with aMore Product Information