Charging stand for one pipette
Supplier: Sartorius
730981
613-4123EA 280 EUR
613-4123
Charging stand for one pipette
Pipette Stands
When the pipette is not in use, it should be stored in an upright position in order to avoid contamination from desks and benches.
The electronic pipettes are recommended to be stored and charged on a charging stand, whenever they are not in use. In this way, the batteries always remain charged, when the work begins. Charging Stand is for storing and charging one electronic pipette, Picus or eLINE®.