Multi-channel pipettes, mechanical, variable volume, mLINE®
Supplier: Sartorius
725120 725130 725140 725220 725230 725240
613-5079EA 1030 EUR
613-5079 613-5080 613-5081 613-5082 613-5083 613-5084
Multi-channel pipettes, mechanical, variable volume, mLINE®
Pipettes Micropipettes
mLINE® pipettes offer a unique combination of new features for higher accuracy and precision. They include built-in replaceable tip-cone filters, which prevent pipette contamination and damage.
  • Clear and easy to read display with big numbers and small increments
  • Equipped with special spring-loaded tip cones (Optiload mechanism) allowing tip loading with equal constant force to every channel using minimum force
  • Plunger technology results in the lowest pipetting force
  • Volume range colour coding for easy pipette selection, and five colour coded caps
  • Thermal