xPIPETTE™ Carousel Stand
Supplier: BIOTIX
30518126
613-6912EA 232 EUR
613-6912
xPIPETTE™ Carousel Stand
Pipette Stands
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
  • Fits both single and multichannel models
  • Holds seven pipettes total
  • Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.