Calibration software, EASYCAL™ 5.0
Supplier: Brand
708410 708414 708412
613-7082EA 1620 EUR
613-7082 613-7084 613-7083
Calibration software, EASYCAL™ 5.0
Software Laboratory Software
The EASYCAL™ 5.0 calibration software facilitates the monitoring of Liquid Handling instruments (like piston-stroke devices such as pipettes, dispensers, burettes, and manual dispensers), as well as volumetric instruments made of glass or plastic, based on GLP/GMP and DIN EN ISO 9001. EASYCAL™ 5 software can be used for BRAND instruments as well as instruments from other manufacturers.
  • Simple: Can be used with pipettes and instruments of all manufacturers
  • Easy: Automatic calculation using the up-to-date standards
  • Efficient: Professional and transparent