VWR® Reagent Reservoirs
Supplier: VWR Collection
613-7284EA 49.5 EUR
613-7284 613-7285 613-7286 613-7287 613-7289 613-7290 613-7291 613-7293 613-7294 613-7297 613-7299 613-7301 613-7304 613-7305 613-7306 613-7307 613-7308 613-7310 613-7311 613-7312 613-7314
VWR® Reagent Reservoirs
Reagent Reservoirs
Reagent reservoirs are designed for efficient liquid transfer, making them an ideal choice for repetitive pipetting tasks. These reagent reservoirs for multichannel pipettes provide a trough wide enough to fit 8 and 12 channel pipettes.
  • Trough design with graduations on the inside wall and pour-off spouts on four corners
  • Slightly tilted inner surface helps reduce residue
  • Non sterile or sterilised options available, achieving SAL 10⁻⁶ (ISO 11137)
  • Reservoirs are lot-certified, non-pyrogenic, DNase- and RNase-free
VWR® reagent reservoirs are an excellentMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M