Precision digital thermometer, TFX 430
Supplier: EBRO
1340-5437 1340-5439 1340-5430 1340-5438 1340-5432
620-0221EA 709 EUR
620-0221 620-0223 620-1385 620-1329 620-2348
Precision digital thermometer, TFX 430
Thermometers Digital Thermometers
This waterproof reference thermometer is suitable for use in a wide range of applications. A microprocessor controls the electronics, guaranteeing maximum measurement precision and consistency over the entire measurement range.
  • Robust and impact resistant
  • 'HOLD' function
  • Automatic switch off
  • Minimum/maximum values displayed
Different PT100 probes for different applications available.
W×D×H: 109×54×22 mm
Weight: 90 g
Delivery information: Supplied with 3-point certificate of calibration, CR2477 3 V lithium battery for 5 years of use. WhenMore Product Information