VWR® Traceable® Platinum Ultra-Accurate Stainless Steel Digital Thermometer
Supplier: VWR Collection
89369-138
620-1980EA 655 EUR
620-1980 VWRU89369-138
VWR® Traceable® Platinum Ultra-Accurate Stainless Steel Digital Thermometer
Thermometers Digital Thermometers
A highly accurate thermometer that is simple to use.
  • Display one, two, or three significant figures after the decimal point
  • Download data to computer for further analysis
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Meter features a splashproof membrane keypad, Memory and Hold functions, an internal data logger that captures the Min/Max temperature every hour for a 24-hour period, and °F/°C selection. The fast-response general-purpose penetration probe (included) displays even the slightest temperatureMore Product Information