Thermometer, mercury, ASTM92C, white backed, measuring range:40:70/0.1C, length:370mm, 76mm immersion, durable pigment, suitable f o r government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items
Supplier: Amarell
Thermometer, mercury, ASTM92C, white backed, measuring range:40:70/0.1C, length:370mm, 76mm immersion, durable pigment, suitable f o r government calibration. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclu 1 * 1 items